3,4,5-Trichlorothiophene-2-carbonyl chloride 97%
Supplier: Thermo Scientific
|
Danger
|
L17704.06
L17704.03
ALFAL17704.06EA
0
SGD
ALFAL17704.06
ALFAL17704.03
3,4,5-Trichlorothiophene-2-carbonyl chloride 97%
3,4,5-Trichlorothiophene-2-carbonyl chloride
Purity: 97%
CAS No.: 24422-15-9
Molecular Formula: C5Cl4OS
Formula Weight: 249.93
Melting Point: 38-40°
Boiling Point: 124-125°/10mm
Beilstein Reference: 1427060
UN No.: UN3261
MDL No.: MFCD02093952
Formula:
C₅Cl₄OS MW: 249,93 g/mol Boiling Pt: 124…125 °C (10 mmHg) Melting Pt: 38…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093952 CAS Number: 24422-15-9 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...