5-tert-Butyl-2-methoxybenzoic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-(tert-Butyl)-2-methoxybenzoic acid
L17833.03
L17833.06
ALFAL17833.03EA
127
SGD
ALFAL17833.03
ALFAL17833.06
5-tert-Butyl-2-methoxybenzoic acid 98%
5-tert-Butyl-2-methoxybenzoic acid
Purity: 98%
CAS No.: 73469-54-2
Molecular Formula: C12H16O3
Formula Weight: 208.26
Melting Point: 74-76°
Beilstein Reference: 2722665
MDL No.: MFCD02093964
Formula:
C₁₂H₁₆O₃ MW: 208,26 g/mol Melting Pt: 74…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093964 CAS Number: 73469-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...