Potassium-4-methyl-β-styryltrifluoroborate 95%

Supplier: Thermo Scientific
Warning

Synonyms: Potassium (E)-trifluoro(4-methylstyryl)borate

L17973.03 L17973.06
ALFAL17973.03EA 187 SGD
ALFAL17973.03 ALFAL17973.06
Potassium-4-methyl-β-styryltrifluoroborate 95%
Potassium-4-methyl-beta-styryltrifluoroborate

Purity: 95% CAS No.: 219718-86-2 Molecular Formula: C9H9BF3K Formula Weight: 224.08 Beilstein Reference: 7783635 MDL No.: MFCD02093982

Formula: C₉H₉BF₃K
MW: 224,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093982
CAS Number: 219718-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR