(S)-(-)-2-Benzylsuccinic acid-1-methyl ester ≥98%, ee 98%

Supplier: Thermo Scientific

Synonyms: (S)-3-Benzyl-4-methoxy-4-oxobutanoic acid

L18265.03 L18265.MD
ALFAL18265.03EA 0 SGD
ALFAL18265.03 ALFAL18265.MD
(S)-(-)-2-Benzylsuccinic acid-1-methyl ester ≥98%, ee 98%
(S)-(-)-2-Benzylsuccinic acid-1-methyl ester

Purity: 98+% CAS No.: 182247-45-6 Molecular Formula: C12H14O4 Formula Weight: 222.24 Beilstein Reference: 7638790 MDL No.: MFCD01311761

Formula: C₁₂H₁₄O₄
MW: 222,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311761
CAS Number: 182247-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR