(S)-(-)-2-Isobutylsuccinic acid 1-methyl ester ≥98%, ee 98%

Supplier: Thermo Scientific

Synonyms: (S)-2-Isobutylsuccinic acid-1-methyl ester

L18268.03 L18268.MD
ALFAL18268.03EA 0 SGD
ALFAL18268.03 ALFAL18268.MD
(S)-(-)-2-Isobutylsuccinic acid 1-methyl ester ≥98%, ee 98%
(S)-(-)-2-Isobutylsuccinic acid 1-methyl ester

Purity: 98+% CAS No.: 213270-36-1 Molecular Formula: C9H16O4 Formula Weight: 188.22 Beilstein Reference: 8110824 MDL No.: MFCD01091033

Formula: C₉H₁₆O₄
MW: 188,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01091033
CAS Number: 213270-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR