(S)-(-)-N-Isopropyl-1-phenylethylamine hydrochloride ≥98%

Supplier: Thermo Scientific
Warning

L18355.06 L18355.03
ALFAL18355.06EA 0 SGD
ALFAL18355.06 ALFAL18355.03
(S)-(-)-N-Isopropyl-1-phenylethylamine hydrochloride ≥98%
(S)-(-)-N-Isopropyl-1-phenylethylamine hydrochloride

Purity: 98+% CAS No.: 116297-12-2 Molecular Formula: C11H17N.HCl Formula Weight: 199.73 Melting Point: 262-265° MDL No.: MFCD06011753

Formula: C₁₁H₁₇N·HCl
MW: 199,73 g/mol
Melting Pt: 262…265 °C
Storage Temperature: Ambient
MDL Number: MFCD06011753
CAS Number: 116297-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR