(3-Chlorophenoxy)acetyl chloride 95%

Supplier: Thermo Scientific
Danger

L18427.06 L18427.03
ALFAL18427.06EA 0 SGD
ALFAL18427.06 ALFAL18427.03
(3-Chlorophenoxy)acetyl chloride 95%
(3-Chlorophenoxy)acetyl chloride

Purity: 95% CAS No.: 114476-84-5 Molecular Formula: C8H6Cl2O2 Formula Weight: 205.04 Flash Point: 110°(230°F) Refractive Index: 1.5480 Boiling Point: 120-122°/2mm Beilstein Reference: 1871611 UN No.: UN3265 MDL No.: MFCD02094018

Formula: C₈H₆Cl₂O₂
MW: 205,04 g/mol
Boiling Pt: 120…122 °C (2 mmHg)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02094018
CAS Number: 114476-84-5
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR