(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine 97%

Supplier: Thermo Scientific
Danger

L18659.03 L18659.MD
ALFAL18659.03EA 840 SGD
ALFAL18659.03 ALFAL18659.MD
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine 97%
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine

Purity: 97% CAS No.: 153837-28-6 Molecular Formula: C18H30N2 Formula Weight: 274.45 Storage Temperature: 2°C to 8°C Melting Point: 36-40° Beilstein Reference: 6957993 Optical Rotation: -90° (c=1 in methanol) UN No.: UN3259 MDL No.: MFCD06795639

Formula: C₁₈H₃₀N₂
MW: 274,45 g/mol
Melting Pt: 36…40 °C
MDL Number: MFCD06795639
CAS Number: 153837-28-6
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR