Bis(neopentyl glycolato)diboron 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5,5,5',5'-Tetramethyl-2,2'-bi(1,3,2-dioxaborinane), 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
L18675.06
L18675.03
L18675.MD
ALFAL18675.06EA
0
SGD
ALFAL18675.06
ALFAL18675.03
ALFAL18675.MD
Bis(neopentyl glycolato)diboron 97%
Bis(neopentyl glycolato)diboron
Purity: 97%
CAS No.: 201733-56-4
Molecular Formula: C10H20B2O4
Formula Weight: 225.89
Storage Temperature: 2°C to 8°C
Melting Point: 170-178°
Beilstein Reference: 8839038
MDL No.: MFCD02093062
Formula:
C₁₀H₂₀B₂O₄ MW: 225,89 g/mol Melting Pt: 170…178 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02093062 CAS Number: 201733-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...