N-(n-Octyl)-n-pentylamine ≥98%

Supplier: Thermo Scientific
Danger

L18869.03 L18869.MD
ALFAL18869.03EA 0 SGD
ALFAL18869.03 ALFAL18869.MD
N-(n-Octyl)-n-pentylamine ≥98%
N-(n-Octyl)-n-pentylamine

Purity: 98+% CAS No.: 6835-13-8 Molecular Formula: C13H29N Formula Weight: 199.38 Refractive Index: 1.4370 Beilstein Reference: 7524567 UN No.: UN2735 MDL No.: MFCD03093075

Formula: C₁₃H₂₉N
MW: 199,38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093075
CAS Number: 6835-13-8
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR