2-Chloro-6-fluoro-3-methylbenzyl alcohol 97%
Supplier: Thermo Scientific
|
Warning
|
L19148.03
L19148.MD
ALFAL19148.03EA
0
SGD
ALFAL19148.03
ALFAL19148.MD
2-Chloro-6-fluoro-3-methylbenzyl alcohol 97%
2-Chloro-6-fluoro-3-methylbenzyl alcohol
Purity: 97%
CAS No.: 261762-83-8
Molecular Formula: C8H8ClFO
Formula Weight: 174.60
Melting Point: 68-70°
MDL No.: MFCD01631362
Formula:
C₈H₈ClFO Melting Pt: 68…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631362 CAS Number: 261762-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...