1,4-Dioxane-2,3-diol ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2,3-Dihydroxy-1,4-dioxane

L19210.14 L19210.06
ALFAL19210.14EA 0 SGD
ALFAL19210.14 ALFAL19210.06
1,4-Dioxane-2,3-diol ≥97%
1,4-Dioxane-2,3-diol

Purity: 97% CAS No.: 4845-50-5 Molecular Formula: C4H8O4 Formula Weight: 120.11 Melting Point: 100-102° Beilstein Reference: 104209 MDL No.: MFCD00006572

Formula: C₄H₈O₄
MW: 120,10 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00006572
CAS Number: 4845-50-5
EINECS: 225-431-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR