5-[4-(Methylthio)phenyl]-1H-tetrazole 99%

Supplier: Thermo Scientific
Warning

L19264.03 L19264.MD
ALFAL19264.03EA 0 SGD
ALFAL19264.03 ALFAL19264.MD
5-[4-(Methylthio)phenyl]-1H-tetrazole 99%
5-[4-(Methylthio)phenyl]-1H-tetrazole

Purity: 99% CAS No.: 138689-79-9 Molecular Formula: C8H8N4S Formula Weight: 192.24 Melting Point: 214-215° MDL No.: MFCD03093097

Formula: C₈H₈N₄S
MW: 192,24 g/mol
Melting Pt: 214…215 °C
Storage Temperature: Ambient
MDL Number: MFCD03093097
CAS Number: 138689-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR