N-Boc-L-isoleucinol 95%

Supplier: Thermo Scientific

Synonyms: tert-Butyl ((2S,3S)-1-hydroxy-3-methylpentan-2-yl)carbamate, Boc-isoleucinol

L19351.03 L19351.06
ALFAL19351.03EA 0 SGD
ALFAL19351.03 ALFAL19351.06
N-Boc-L-isoleucinol 95%
N-Boc-L-isoleucinol

Purity: 95% CAS No.: 106946-74-1 Molecular Formula: C11H23NO3 Formula Weight: 217.31 Storage Temperature: 2°C to 8°C Refractive Index: 1.4530 Boiling Point: 121-126°/0.6mm Beilstein Reference: 3604099 MDL No.: MFCD00235929

Formula: C₁₁H₂₃NO₃
MW: 217,3 g/mol
Boiling Pt: 121…126 °C (0,6 mmHg)
MDL Number: MFCD00235929
CAS Number: 106946-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR