(S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine ≥85%, tech.
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(S)-Methoxymethyl-(1-phenyl-ethyl)-trimethylsilanylmethyl-amine
L19414.03
L19414.06
ALFAL19414.03EA
0
SGD
ALFAL19414.03
ALFAL19414.06
(S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine ≥85%, tech.
(S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine
Purity: 85%
CAS No.: 143140-08-3
Molecular Formula: C14H25NOSi
Formula Weight: 251.45
Boiling Point: 82-84°/0.2mm
Beilstein Reference: 5906612
Optical Rotation: -47° (c=1 in carbon tetrachloride)
MDL No.: MFCD06798960
Formula:
C₁₄H₂₅NOSi MW: 251,45 g/mol Boiling Pt: 82…84 °C (0,2 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD06798960 CAS Number: 143140-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...