Boc-cycloleucine 98%
Supplier: Thermo Scientific
Synonyms:
1-(Boc-amino)cyclopentanecarboxylic acid, 1-N-BOC-Aminocyclopentanecarboxylic acid, 1-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
L19483.03
L19483.06
ALFAL19483.03EA
108
SGD
ALFAL19483.03
ALFAL19483.06
Boc-cycloleucine 98%
Boc-cycloleucine
Purity: 98%
CAS No.: 35264-09-6
Molecular Formula: C11H19NO4
Formula Weight: 229.28
Melting Point: 131-134°
Beilstein Reference: 2734412
MDL No.: MFCD01076126
Formula:
C₁₁H₁₉NO₄ MW: 229,28 g/mol Melting Pt: 131…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076126 CAS Number: 35264-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...