2,3-Dimercapto-1-propanesulphonic acid sodium salt monohydrate 95%
Supplier: Thermo Scientific
L19519.03
L19519.MD
ALFAL19519.03EA
0
SGD
ALFAL19519.03
ALFAL19519.MD
2,3-Dimercapto-1-propanesulphonic acid sodium salt monohydrate 95%
2,3-Dimercapto-1-propanesulphonic acids odium salt monohydrate
Purity: 95%
CAS No.: 207233-91-8
Molecular Formula: C3H7NaO3S3.H2O
Formula Weight: 228.30 (210.28anhy)
Melting Point: ca 229° dec.
Beilstein Reference: 3734863
MDL No.: MFCD00149543
Formula:
C₃H₉NaO₄S₃·H₂O Melting Pt: ∼ 229 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149543 CAS Number: 207233-91-8 EINECS: 223-796-3 Merck Index: 14,3210 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...