(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥99%, ee 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(3R)-(+)-3-(Boc-amino)pyrrolidine, (R)-tert-Butyl pyrrolidin-3-ylcarbamate
L19692.06
L19692.03
ALFAL19692.06EA
0
SGD
ALFAL19692.06
ALFAL19692.03
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥99%, ee 99%
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine
Purity: 99%
CAS No.: 122536-77-0
Molecular Formula: C9H18N2O2
Formula Weight: 186.25
Storage Temperature: 2°C to 8°C
Melting Point: 50-54°
Beilstein Reference: 5377812
Optical Rotation: +21° (c=1 in ethanol)
UN No.: UN3259
MDL No.: MFCD00143191
Formula:
C₉H₁₈N₂O₂ MW: 186,26 g/mol Melting Pt: 50…54 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00143191 CAS Number: 122536-77-0 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...