3,3-Difluoropiperidine hydrochloride 99%
Supplier: Thermo Scientific
|
Warning
|
L19777.MD
L19777.03
ALFAL19777.MDEA
0
SGD
ALFAL19777.MD
ALFAL19777.03
3,3-Difluoropiperidine hydrochloride 99%
3,3-Difluoropiperidine hydrochloride
Purity: 99%
CAS No.: 496807-97-7
Molecular Formula: C5H9F2N.HCl
Formula Weight: 157.59
Melting Point: ca 240°
MDL No.: MFCD03452800
Formula:
C5H10ClF2N MW: 157,59 g/mol Melting Pt: ∼ 240 °C Storage Temperature: Ambient |
MDL Number:
MFCD03452800 CAS Number: 496807-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...