1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal 98%
Supplier: Thermo Scientific
Synonyms:
8-Amino-1,4-dioxaspiro[4.5]decane-8-carboxylic Acid
L19795.03
L19795.MD
ALFAL19795.03EA
406
SGD
ALFAL19795.03
ALFAL19795.MD
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal 98%
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal
Purity: 98%
CAS No.: 54621-18-0
Molecular Formula: C9H15NO4
Formula Weight: 201.22
Storage Temperature: 2°C to 8°C
Melting Point: 301-304°
Beilstein Reference: 1373325
MDL No.: MFCD03790906
Formula:
C₉H₁₅NO₄ MW: 201,22 g/mol Melting Pt: 301…304 °C |
MDL Number:
MFCD03790906 CAS Number: 54621-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...