4-(Cyanomethyl)benzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetonitrile, 4-(Cyanomethyl)phenylboronic acid pinacol ester

L19956.03 L19956.06
ALFAL19956.03EA 0 SGD
ALFAL19956.03 ALFAL19956.06
4-(Cyanomethyl)benzeneboronic acid pinacol ester 95%
4-(Cyanomethyl)benzeneboronic acid pinacol ester

Purity: 95% CAS No.: 138500-86-4 Molecular Formula: C14H18BNO2 Formula Weight: 243.11 Melting Point: 74-78° MDL No.: MFCD02179459

Formula: C₁₄H₁₈BNO₂
MW: 243,11 g/mol
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD02179459
CAS Number: 138500-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR