2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine 98%
Supplier: Thermo Scientific
|
Danger
|
L20026.03
L20026.MD
ALFAL20026.03EA
0
SGD
ALFAL20026.03
ALFAL20026.MD
2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine 98%
2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine
Purity: 98%
CAS No.: 628692-22-8
Molecular Formula: C6H2ClF4N
Formula Weight: 199.54
Refractive Index: 1.4400
Boiling Point: 35-37°/25mm
UN No.: UN2810
MDL No.: MFCD04038326
Formula:
C₆H₂ClF₄N Boiling Pt: 35…37 °C (25 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD04038326 CAS Number: 628692-22-8 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...