1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-methylbenzene 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Methyl-2,3,5,6-tetrafluorobenzyl bromide
L20210.03
L20210.06
ALFAL20210.03EA
0
SGD
ALFAL20210.03
ALFAL20210.06
1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-methylbenzene 98%
1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-methylbenzene
Purity: 98%
CAS No.: 92814-00-1
Molecular Formula: C8H5BrF4
Formula Weight: 257.03
Melting Point: 41-43°
Boiling Point: 49°/0.4mm
Beilstein Reference: 5003900
UN No.: UN3261
MDL No.: MFCD03001155
Formula:
C₈H₅BrF₄ MW: 257,03 g/mol Boiling Pt: 49 °C (0,4 mmHg) Melting Pt: 41…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD03001155 CAS Number: 92814-00-1 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...