3,4-Dimethoxyphenylglyoxal hydrate 98% (dry weight)
Supplier: Thermo Scientific
|
Warning
|
L20279.03
L20279.06
ALFAL20279.03EA
165
SGD
ALFAL20279.03
ALFAL20279.06
3,4-Dimethoxyphenylglyoxal hydrate 98% (dry weight)
3,4-Dimethoxyphenylglyoxal hydrate
Purity: 98%
CAS No.: 163428-90-8
Molecular Formula: C10H10O4.xH2O
Formula Weight: 194.19(anhy)
Melting Point: 118-120°
Beilstein Reference: 3092619
MDL No.: MFCD06800598
Formula:
C₁₀H₁₀O₄ xH₂O MW: 194,19 g/mol (anhy) Melting Pt: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD06800598 CAS Number: 163428-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...