5-tert-Butyl-2-methoxybenzeneboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(5-(tert-Butyl)-2-methoxyphenyl)boronic acid, 5-(tert-Butyl)-2-methoxyphenylboronic acid
L20415.03
L20415.MD
ALFAL20415.03EA
256
SGD
ALFAL20415.03
ALFAL20415.MD
5-tert-Butyl-2-methoxybenzeneboronic acid ≥98%
5-tert-Butyl-2-methoxybenzeneboronic acid
Purity: 98+%
CAS No.: 128733-85-7
Molecular Formula: C11H17BO3
Formula Weight: 208.07
Melting Point: 103-106°
Beilstein Reference: 4181890
MDL No.: MFCD06201034
Formula:
C₁₁H₁₇BO₃ MW: 208,07 g/mol Melting Pt: 103…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD06201034 CAS Number: 128733-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...