Dimenhydrinate, powder
Supplier: MP Biomedicals
Synonyms:
Amosyt, 8-Chloro-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione 2-(benzhydryloxy)-N,N-dimethylethanamine salt
0219018925
0219018910
ICNA0219018925EA
318
SGD
ICNA0219018925
ICNA0219018910
Dimenhydrinate, powder
Dimenhydrinate
Soluble in ethanol (50 mg/mL) giving clear solution.
Formula:
C₁₇H₂₁NO·C₇H₇ClN₄O₂ MW: 469,96 g/mol |
MDL Number:
MFCD00054265 CAS Number: 523-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...