HPLC reservoirs, single cavity, PYREX®
Supplier: Corning
2150-1L 2150-2L
CORN2150-1LEA 433 SGD
CORN2150-1L CORN2150-2L
HPLC reservoirs, single cavity, PYREX®
Bottles HPLC Bottles
These reservoirs have a central cavity for a mobile phase inlet reservoir, an autoclavable plastic coat that absorbs UV up to 385 nm and a GL 45 thread finish.