Inulin (from chicory root), free flowing powder

Supplier: MP Biomedicals

Synonyms: Dahlin, Alantin, Inulin (by Enzymatic synthesis)

0219897110 0219897125 0219897180 0219897191
ICNA0219897110EA 105 SGD
ICNA0219897110 ICNA0219897125 ICNA0219897180 ICNA0219897191
Inulin (from chicory root), free flowing powder
Inulin

A heterogeneous blend of fructose polymers that function as plant storage carbohydrates.


Inulin are potential replacement for high fructose corn syrup due to its prebiotic function and similar structural characteristics to corn syrup.


Inulin are potential replacement for high fructose corn syrup due to its prebiotic function and similar structural characteristics to corn syrup.


A heterogeneous blend of fructose polymers that function as plant storage carbohydrates.


Soluble in hot water; slightly soluble in cold water and organic solvents.

Melting Pt: 158…165 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00131407
CAS Number: 9005-80-5

Order Now

Specification Test Results

Identity Test Passes
Source Chicory Root
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR