Norethindrone, white powder

Supplier: MP Biomedicals
Danger

Synonyms: Norethisterone

0215594691 0215594694
ICNA0215594691EA 301 SGD
ICNA0215594691 ICNA0215594694
Norethindrone, white powder
Norethindrone

Norethindrone is a molecule used in some combined oral contraceptive pills, progestogen only pills and is also available as a stand-alone drug.


It is a progestogen and can be used to treat premenstrual syndrome, painful periods, abnormal heavy bleeding, irregular periods, menopausal syndrome (in combination with oestrogen), or to postpone a period. It is also commonly used to help prevent uterine hemorrhage in complicated non-surgical or pre-surgical gynecologic cases.


Norethindrone is a synthetic progestin that possesses antiestrogenic effects, where in low doses it is progestational and in high doses it is antiestrogenic. It also causes a time-dependent loss of cytochrome P-450 when incubated in vitro with rat liver microsomal fractions and NADPH-generating systems.


Soluble in chloroform: ≥50 mg/ml, clear, colorless.

Formula: C₂₀H₂₆O₂
MW: 298,426 g/mol
Melting Pt: 203,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00067596
CAS Number: 68-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR