Hyaluronic acid (from bovine vitreous humor), white powder

Supplier: MP Biomedicals

0215993310 0215993350
ICNA0215993310EA 568 SGD
ICNA0215993310 ICNA0215993350
Hyaluronic acid (from bovine vitreous humor), white powder
Hyaluronic acid

A high viscosity mucopolysaccharide composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine which forms the core of complex proteoglycan aggregates found in extracellular matrix.


Hyaluronic Acid is used in study of activation of the MAPK ERK following Leishmania amazonensis, infection of Macrophages.


High molecular mass polymer composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine which forms the core of complex proteoglycan aggregates found in extracellular matrix.


Dissolves at 10 mg/mL in analytical grade water to give a clear, viscous solution, after soaking overnight at +5 °C then stirring at room temperature for approximately 1 hour.

Formula: C₂₈H₄₄N₂O₂₃
Storage Temperature: Refrigerator
MDL Number: MFCD00131348
CAS Number: 9004-61-9

Order Now

Specification Test Results

Identity Test Passes
Source Bovine Vitreous Humor

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR