Goat serum
Supplier: MP Biomedicals
0219135680 0219135691
ICNA0219135680EA 146 SGD
ICNA0219135680 ICNA0219135691
Goat serum
Sera (Serum)
Goat serum is a pooled preparation obtained from a normal donor herd.
Serum, goat is suitable replacement for FBS on most cell lines and primary cultures for biomedical research - virus isolation and identification, virological studies, etc. It is used as an effective NBCS replacement for the culture of fish cells and could be successfully used in place of fetal bovine serum in the growth medium for long-term in vitro propagation of T. annulata schizonts. This is used for blocking agents and controls in immunoassays.