Rabbit serum
Supplier: MP Biomedicals
0219135720 0219135780
ICNA0219135720EA 48.8 SGD
ICNA0219135720 ICNA0219135780
Rabbit serum
Sera (Serum)
Rabbit serum is a component of blood obtained from a rabbit that contains blood plasma without fibrinogen or other clotting factors.
Rabbit serum combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry. In immunohistochemistry, antibodies are used to detect the presence and abundance of certain antigens in a given tissue sample. The use of a blocking serum such as rabbit serum is important because it prevents nonspecific binding and interaction between the antibodies and non-targeted antigens. Rabbit serum is also a common component in media used to maintain cell culture.