Stirrer bearings, PTFE
Supplier: CHEMGLASS INC
CG-2077-03 CG-2077-20 CG-2077-21
CGLACG-2077-03EA 1480 SGD
CGLACG-2077-03 CGLACG-2077-20 CGLACG-2077-21
Stirrer bearings, PTFE
Stirrers Stirring Tools
These grease-free PTFE stirrer bearings consist of a threaded glass adapter with the listed inner joint, a saddle o-ring, a body bushing with a compression and locking nut, and a precision PTFE inner bearing.
  • Ideal for use in moderate vacuum applications
  • Saddle o-ring allows only glass and PTFE contact with the reaction, while ensuring a vacuum tight seal