Screw caps, DURAN® PURE Premium STERICLIN®-Packed, DURAN®
Supplier: DWK Life Sciences
892992806 892991304
SCOT892992806EA 575 SGD
SCOT892992806 SCOT892991304
Screw caps, DURAN® PURE Premium STERICLIN®-Packed, DURAN®
Closures Bottle Closures
STERICLIN®-packaging offers a sterile barrier system (SBS).
  • No pre-cleaning of the Premium cap is necessary
  • Ideal for dry heat sterilisation / depyrogenation
  • Can be sterilised directly in the STERICLIN® pouches
  • Sterile storage for up to 6 months
  • Pouch contains caps from a single manufacturing lot
These paper/film see-through pouches are compliant in accordance with DIN ISO 11607. The sterilisation-proof paper is an effective barrier against germs and bacteria, yet it permits the transmission of air, steam, or sterilisation gases. It‘s purpose is toMore Product Information