TLC spotting capillary tubes
Supplier: CHEMGLASS
CG-1182-01 CG-1182-02
CGLACG-1182-01EA 467 SGD
CGLACG-1182-01 CGLACG-1182-02
TLC spotting capillary tubes
Pipettes Microcapillaries
Non-calibrated tubes made from borosilicate glass for use with aqueous and organic solutions.
  • Very good temperature resistance
  • Very good chemical resistance