Powder funnels
Supplier: CHEMGLASS INC
CG-1720-01 CG-1720-02
CGLACG-1720-01EA 110 SGD
CGLACG-1720-01 CGLACG-1720-02
Powder funnels
Funnels Filtration Funnels Funnels with Conical Joints
Funnels have 60° side angle and a standard ground joint.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.