Guinea pig serum
Supplier: MP Biomedicals
08642831 08642836
ICNA08642831EA 321 SGD
ICNA08642831 ICNA08642836
Guinea pig serum
Sera (Serum)
Guinea pig serum is rapidly collected and processed under cold room conditions which maintains maximum complement activity.
  • Suitable for use in hemolytic titrations and cytotoxicity assays
  • Suitable for use as a serum protein control in immunoassays