Adapters, thermometer, inlet
Supplier: CHEMGLASS
CG-1042-01 CG-1042-14
CGLACG-1042-01EA 118 SGD
CGLACG-1042-01 CGLACG-1042-14
Adapters, thermometer, inlet
Adapters Adapters, Joints with Tube Connectors
Adapters have a No.7 chem-thread at at the top for vacuum tight seal of plain stem thermometers or any other tube having an O.D. between 4 and 7 mm.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Delivery information: Supplied complete with a Celcon® compression cap and Viton o-ring.