H-Lys(Fmoc)-OH ≥98%

Supplier: Thermo Scientific

Synonyms: Nepsilon-Fmoc-L-lysine

H62834.03 H62834.06
ALFAH62834.03EA 133 SGD
ALFAH62834.03 ALFAH62834.06
H-Lys(Fmoc)-OH ≥98%
H-Lys(Fmoc)-OH

N^e-Fmoc-L-lysine, 97%


Purity: 97% CAS No.: 84624-28-2 Molecular Formula: C21H24N2O4 Formula Weight: 368.43 MDL No.: MFCD00038365

Formula: C₂₁H₂₄N₂O₄
MW: 368,433 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038365
CAS Number: 84624-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR