Yeast aldehyde dehydrogenase, MP Biomedicals
Supplier: MP Biomedicals
0219067925 0219067980 0219067983
ICNA0219067925EA 146 SGD
ICNA0219067925 ICNA0219067980 ICNA0219067983
Yeast aldehyde dehydrogenase, MP Biomedicals
Enzymes
  • From Yeast Potassium Activated (Aldehyde: NAD + (P+) ov. reductase)
  • E.C.1.2.1.5
  • Freeze dried stabilized powder
  • Activity: 2 unit/mg solid (approx. 5 u/mg protein)
  • ADH Less than 0,1% Unit
Definition: One unit causes the oxidation of one micromole of acetaldehyde to acetic acid per minute at pH 8,0 and 25 °C in the presence of potassium ions and thiols.