β-Pseudouridine (synthetic) (by HPLC)

Supplier: MP Biomedicals

Synonyms: Pseudouridine, 5-((2S,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

0219917850 0219917810 0219917883
ICNA0219917850EA 2250 SGD
ICNA0219917850 ICNA0219917810 ICNA0219917883
β-Pseudouridine (synthetic) (by HPLC)
β-Pseudouridine

β-Pseudouridine isomer of the nucleoside uridine found in all species and in many classes of RNA except mRNA. It is formed by enzymes called Ψ synthases, which post-transcriptionally isomerize specific uridine residues in RNA in a process termed pseudouridylation. Studies suggest that β-Pseudouridine reduces radiation-induced chromosome aberrations in human lymphocytes.
+4 °C


  • Presentation: Off-White Powder
  • Soluble in water (50 mg/mL - clear, colorless solution).


Storage temperature.: +4 °C

Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Boiling Pt: 598.4 °C (760 mmHg)) (lit.)
Density: 1.64 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00038458
CAS Number: 1445-07-4

Order Now

Specification Test Results

Identity Test Passes
Source Synthetic

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR