Volumetric flasks, class A, with screw cap, EZ Access™, PYREX®
Supplier: Corning
5655-100 5655-1L 5655-200 5655-250 5655-50 5655-500
CORN5655-100EA 100 SGD
CORN5655-100 CORN5655-1L CORN5655-200 CORN5655-250 CORN5655-50 CORN5655-500
Volumetric flasks, class A, with screw cap, EZ Access™, PYREX®
Flasks Volumetric Flasks
These PYREX® EZ Access™ heavy walled volumetric flasks are designed to meet Class A, ASTM tolerances for volumetrics.
  • Calibrated 'to contain' (TC)
  • GL 32 high temperature screw cap fits all flasks
  • Screw cap with PTFE liner provides a secure closure for leak proof storage of solutions
  • Extra large volume size is permanently screened onto the flask body for easy volume identification
The wide neck design allows for easy filling and mixing of various materials into solution and also allows access to larger capacity pipettes.
Order Now
SPECIFICATIONS
Environmentally Preferable