Moisture test distilling receiver, PYREX®
Supplier: Corning
3611-10 3611-25
CORN3611-10EA 165 SGD
CORN3611-10 CORN3611-25
Moisture test distilling receiver, PYREX®
Distillation Components Distillation Receivers
Receivers, made of PYREX® brosilicate glass, are designed for determining water by distillation.
  • Have 24/40 standard taper ground joint on the sidearm
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The 10 ml receiver is graduated from 0 to 1 ml in 0,1 ml increments, with a maximum scale error of 0,05 ml. The graduation intervals from 1,0 to 10,0 ml are 0,2 ml, with maximum scale error of 0,1 ml. The 25 ml receiver is graduated from 0 ml to 1 ml, in 0,1 ml increments, with a maximum scale error of 0,05 ml. The graduation intervals from 1,0 to 25,0 ml are 0,2 ml, with a maximum scale error of 0,1 ml.
Certifications: ASTM E-123.