West condenser, PYREX®
Supplier: Corning
2705-19 2800-300
CORN2705-19EA 825 SGD
CORN2705-19 CORN2800-300
West condenser, PYREX®
Condensers
Made of PYREX® borosilicate glass.
  • Tubulations on the same side reduce breakage
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This West condenser is designed with a long heavy-wall outer jacket to provide a sturdy, long-lasting unit, while the inner tube has a thinner wall for efficient heat transfer.
It has a standard taper outer joint at the top and a standard taper inner joint at the bottom with a drip tip. The tubulation Ext.Ø is approximately 10 mm.