Erlenmeyer flasks, narrow neck, with baffles, PYREX®
Supplier: Corning
4450-125 4450-1L 4450-250
CORN4450-125EA 257 SGD
CORN4450-125 CORN4450-1L CORN4450-250
Erlenmeyer flasks, narrow neck, with baffles, PYREX®
Flasks Erlenmeyer Flasks
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • For convenience, these flasks include a marking spot
  • Uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance
Erlenmeyer flasks with heavy tooled rims for maximum durability. Also the baffle design promotes optimal and consistent solution agitation when used with flask shakers.