Serological pipettes, for cotton plugging, PYREX®
Supplier: Corning
7086-10 7086-5
CORN7086-10EA 191 SGD
CORN7086-10 CORN7086-5
Serological pipettes, for cotton plugging, PYREX®
Pipettes Serological Pipettes
Glass, reusable. These serological pipettes are designed for cotton plugging. The uniform top-end openings permit performing of plugs and allow precise control of liquid column.
  • Colour coded by size for easy identification and sorting
  • Coloured graduations are enameled onto the glass
  • Calibrated to deliver (TD)
Graduation interval: 0,1 ml.
Certifications: ASTM E-1044
Order Now
SPECIFICATIONS
Environmentally Preferable