PYREX® Filter Tube Funnels
Supplier: Corning
9480-24 9480-27 9480-32 9480-36 9480-42
CORN9480-24EA 19.6 SGD
CORN9480-24 CORN9480-27 CORN9480-32 CORN9480-36 CORN9480-42
PYREX® Filter Tube Funnels
Funnels Filtration Funnels Filter Funnels
Made from PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Filter tubes have a constricted lower end. They can be used on the bottom end of 30 mm O.D. PYREX® Gooch low form fritted disc crucibles to collect the filtrate. They can also be used directly with porcelain plates.
The constricted lower end is approximately 75 mm long by 8 mm O.D. on all sides.