Kuderna-Danish concentrator tubes, jacketed, PYREX®
Supplier: Corning
2158-10JTO 2158-1JTO
CORN2158-10JTOEA 212 SGD
CORN2158-10JTO CORN2158-1JTO
Kuderna-Danish concentrator tubes, jacketed, PYREX®
Concentrators
These innovative jacketed Kuderna-Danish concentrator tubes circulate hot water through the jacket to boil the solvent during extraction/concentration.
  • Bottom portion of the tube is never heated, so samples won't boil dry
  • Concentration automatically stops when the solvent sinks below the jacket
  • Lab technician time is saved by eliminating the need to watch evaporations closely to avoid boiling dry
  • Serrated tubulations allow for easy tubing connections
Standard taper top joint: 19/22.
Order Now
SPECIFICATIONS
Environmentally Preferable