Ignition tubes, without rim, heavy wall, PYREX®
Supplier: Corning
9860-14 9860-16 9860-25
CORN9860-14EA 739 SGD
CORN9860-14 CORN9860-16 CORN9860-25
Ignition tubes, without rim, heavy wall, PYREX®
Tubes Test Tubes
These PYREX® rimless ignition tubes are made from No. 7740 glass.
  • Very good chemical resistance
  • Good temperature resistance
These ignition tubes are satisfactory for most applications encountered in school work where temperatures above 600 °C are not required. These rimless tubes perform well when continually used at temperatures up to 500 °C, or for short periods to 600 °C.