Volumetric pipettes, class A, serialised, PYREX®
Supplier: Corning
7103C-1 7103C-10 7103C-15 7103C-2 7103C-20 7103C-25 7103C-3 7103C-4 7103C-5 7103C-6 7103C-7 7103C-8 7103C-9
CORN7103C-1EA 141 SGD
CORN7103C-1 CORN7103C-10 CORN7103C-15 CORN7103C-2 CORN7103C-20 CORN7103C-25 CORN7103C-3 CORN7103C-4 CORN7103C-5 CORN7103C-6 CORN7103C-7 CORN7103C-8 CORN7103C-9
Volumetric pipettes, class A, serialised, PYREX®
Pipettes Graduated Pipettes
These PYREX® volumetric pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
  • Reusable precision pipettes
  • Colour-coded markings are enameled onto the glass
PYREX® serialised/certified volumetric glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Each pipette is individually serialized and supplied with a Certificate of Identification and capacity, traceable to NIST standards. These precision PYREX® volumetric pipettes are manufactured and calibrated 'To Contain' (TC: bottom line)More Product Information